Dimethyl-5-bromoisophthalate 97%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 5-bromoisophthalate, 5-Bromoisophthalic acid dimethyl ester

A228340-100G A228340-10G A228340-500G A228340-1G A228340-25G A228340-5G
103819-390EA 9 USD
103819-390 103850-298 76771-060 76771-062 76771-064 77089-082 77089-084 77089-086
Dimethyl-5-bromoisophthalate 97%
Dimethyl-5-bromoisophthalate

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₄
MW: 273.08 g/mol
Melting Pt: 85…89 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00078709
CAS Number: 51760-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR