3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzenedicarboxylic acid dimethyl ester, Dimethyl 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalate, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalic acid dimethyl ester, 3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester, Dimethyl 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzenedicarboxylate, 2-[3,5-Bis(methoxycarbonyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A207573-1G A207573-5G A207573-100MG A207573-250MG
103817-806EA 16 USD
103817-806 103848-488 77086-772 77086-774 77164-840 77164-842
3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester 98%
3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₁BO₆
MW: 320.148 g/mol
Boiling Pt: 435.4±35.0 °C (760 mm Hg)
Melting Pt: 126…127 °C
Flash Pt: 217.1 °C
Storage Temperature: Ambient
MDL Number: MFCD11858596
CAS Number: 944392-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR