Dimethyl 4-hydroxyphthalate 98%

Supplier: Ambeed
Warning

A100419-100G A100419-100MG A100419-10G A100419-250MG A100419-25G A100419-1G A100419-5G
103795-426EA 8 USD
103795-426 103821-122 77093-466 77093-468 77093-470 77093-472 77093-474 77296-674 77296-676
Dimethyl 4-hydroxyphthalate 98%
Dimethyl 4-hydroxyphthalate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₅
MW: 210.18 g/mol
Boiling Pt: 330.6 °C (760 mm Hg)
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00060092
CAS Number: 22479-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR