Dimethyl-4-chloropyridine-2,6-dicarboxylate 98%

Supplier: Ambeed
Warning

A259712-10G A259712-250MG A259712-1G A259712-25G A259712-5G A259712-100G
103812-460EA 8 USD
103812-460 103852-342 76815-992 76815-994 77091-922 77091-924 77091-926 77883-713
Dimethyl-4-chloropyridine-2,6-dicarboxylate 98%
Dimethyl-4-chloropyridine-2,6-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₈ClNO₄
Boiling Pt: 355.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02612781
CAS Number: 5371-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR