Dimethyl 4-bromopyridine-2,6-dicarboxylate 97%

Supplier: Ambeed
Warning

A462498-100MG A462498-1G A462498-250MG
103827-940EA 43 USD
103827-940 103839-016 76804-656 76842-904 77602-010
Dimethyl 4-bromopyridine-2,6-dicarboxylate 97%
Dimethyl 4-bromopyridine-2,6-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₈BrNO₄
MW: 274.07 g/mol
Boiling Pt: 369.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09910266
CAS Number: 162102-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR