Dimethyl 4-bromophthalate 98%

Supplier: Ambeed
Warning

A314791-1G A314791-25G A314791-100G A314791-10G A314791-5G
103825-046EA 13 USD
103825-046 103835-970 76838-790 76838-792 77114-658 77114-660 77306-120
Dimethyl 4-bromophthalate 98%
Dimethyl 4-bromophthalate

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₄
Boiling Pt: 305.1 °C (760 mm Hg)
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD07772010
CAS Number: 87639-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR