Dimethyl-4,5-imidazoledicarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 1H-Imidazole-4,5-dicarboxylate, Dimethyl 4,5-imidazoledicarboxylate, 1H-Imidazole-4,5-dicarboxylic acid dimethyl ester

A344282-100G A344282-10G A344282-1G A344282-250MG A344282-25G A344282-5G
103836-878EA 12 USD
103836-878 76773-204 76773-206 76773-208 76773-210 76839-924 76839-926
Dimethyl-4,5-imidazoledicarboxylate 98%
Dimethyl-4,5-imidazoledicarboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₄
MW: 184.1500 g/mol
Boiling Pt: 338.2 °C (760 mm Hg)
Melting Pt: 197…201 °C
Flash Pt: 158.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00051338
CAS Number: 3304-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR