Dimethyl 3-chloropyridine-2,5-dicarboxylate 96%

Supplier: Ambeed
Warning

A499684-250MG A499684-1G A499684-5G
103828-282EA 103 USD
103828-282 103839-370 76830-160 77307-612 77307-614
Dimethyl 3-chloropyridine-2,5-dicarboxylate 96%
Dimethyl 3-chloropyridine-2,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₈ClNO₄
MW: 229.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129113
CAS Number: 106014-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR