Dimethyl 3-aminophthalate 98%

Supplier: Ambeed
Warning

A268629-1G A268629-25G A268629-5G
103812-924EA 12 USD
103812-924 103852-826 77092-582 77092-584 77092-586
Dimethyl 3-aminophthalate 98%
Dimethyl 3-aminophthalate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
Boiling Pt: 306 °C (760 mm Hg)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD09029586
CAS Number: 34529-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR