Dimethyl 3-aminopentanedioate acetate 97%
Supplier: Ambeed
|
Warning
|
A117181-100MG
A117181-1G
103798-842EA
279
USD
103798-842
103824-714
77009-236
77297-518
Dimethyl 3-aminopentanedioate acetate 97%
Dimethyl 3-aminopentanedioate acetate
Precaución: for Research Use Only
Formula:
C₉H₁₇NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD27952975 CAS Number: 1345983-89-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...