Dimethyl 2-propylmalonate 97%
Supplier: Ambeed
|
Warning
|
A170818-10G
A170818-1G
A170818-5G
A170818-25G
103806-546EA
12
USD
103806-546
103844-822
76799-388
77081-942
77081-944
77301-080
Dimethyl 2-propylmalonate 97%
Dimethyl 2-propylmalonate
Precaución: for Research Use Only
Formula:
C₈H₁₄O₄ Boiling Pt: 204.4 °C (760 mm Hg) Flash Pt: 90.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07778475 CAS Number: 14035-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...