Dimethyl chloromalonate 90%
Supplier: Ambeed
|
Danger
|
Synonyms:
Chloromalonic acid dimethyl ester, Dimethyl 2-chloromalonate
A259357-100G
A259357-10G
A259357-1G
A259357-25G
A259357-5G
103812-436EA
7
USD
103812-436
103852-316
76801-746
76801-748
77091-868
77091-870
77091-872
Dimethyl chloromalonate 90%
Dimethyl chloromalonate
Precaución: for Research Use Only
Formula:
C₅H₇ClO₄ Boiling Pt: 215 °C (760 mm Hg) Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042837 CAS Number: 28868-76-0 UN: 3265 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...