Dimethyl 2-bromoisophthalate 95%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 2-bromobenzene-1,3-dicarboxylate, 2-bromoisophthalic acid methyl ester

A121337-100MG A121337-250MG A121337-1G A121337-5G
103800-060EA 39 USD
103800-060 77009-442 77009-444 77297-824 77297-826
Dimethyl 2-bromoisophthalate 95%
Dimethyl 2-bromoisophthalate

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₄
Boiling Pt: 313.0±22.0 °C (760 mm Hg)
Flash Pt: 143.1 °C
Storage Temperature: Ambient
MDL Number: MFCD17169956
CAS Number: 39622-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR