Dimethyl 2-aminoisophthalate 98%

Supplier: Ambeed
Warning

A236506-5G A236506-1G A236506-250MG
103811-104EA 18 USD
103811-104 103850-876 77089-926 77165-678 77304-052 77892-405
Dimethyl 2-aminoisophthalate 98%
Dimethyl 2-aminoisophthalate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD18431747
CAS Number: 57053-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR