Dimethyl 2,5-dibromoterephthalate 97%
Supplier: Ambeed
|
Warning
|
A147157-250MG
A147157-1G
A147157-25G
A147157-5G
A147157-100G
A147157-10G
103803-744EA
7
USD
103803-744
103841-840
76798-468
77317-200
77317-202
77317-204
77879-031
77879-032
Dimethyl 2,5-dibromoterephthalate 97%
Dimethyl 2,5-dibromoterephthalate
Precaución: for Research Use Only
Formula:
C₁₀H₈Br₂O₄ Boiling Pt: 362 °C (760 mm Hg) Flash Pt: 172.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00204592 CAS Number: 18014-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...