Dimethyl iminodiacetate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 2,2'-azanediyldiacetate hydrochloride

A147944-100G A147944-10G A147944-1G A147944-25G A147944-5G
103803-852EA 6 USD
103803-852 103841-968 77162-624 77162-626 77317-388 77317-390 77317-392
Dimethyl iminodiacetate hydrochloride 98%
Dimethyl iminodiacetate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₄·HCl
MW: 197.62 g/mol
Boiling Pt: 210.3 °C (760 mm Hg)
Melting Pt: 170 °C
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216724
CAS Number: 39987-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR