1-Methyl-1H-pyrazole-3,5-dicarboxylic acid dimethyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 1-methyl-1H-pyrazole-3,5-dicarboxylate

A141914-10G A141914-250MG A141914-1G A141914-25G A141914-5G
103803-078EA 9 USD
103803-078 77190-486 77190-488 77316-280 77316-282 77316-284
1-Methyl-1H-pyrazole-3,5-dicarboxylic acid dimethyl ester 97%
1-Methyl-1H-pyrazole-3,5-dicarboxylic acid dimethyl ester

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₄
MW: 198.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443806
CAS Number: 33146-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR