Dimethyl biphenyl-4,4'-dicarboxylate 98%
Supplier: Ambeed
|
Warning
|
A384879-5G
A384879-100G
A384879-10G
A384879-500G
A384879-1G
A384879-25G
103826-772EA
9
USD
103826-772
103837-810
76841-154
77033-466
77033-468
77033-470
77306-962
77306-964
Dimethyl biphenyl-4,4'-dicarboxylate 98%
Dimethyl biphenyl-4,4'-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₆H₁₄O₄ Boiling Pt: 407 °C (760 mm Hg) Melting Pt: 210…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017201 CAS Number: 792-74-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...