Dimethyl (hydroxymethyl)phosphonate 91%

Supplier: Ambeed
Warning

A320317-100G A320317-10G A320317-250MG A320317-1G A320317-5G
103825-232EA 11 USD
103825-232 103836-166 76828-176 76828-178 76828-180 76839-038 76839-040
Dimethyl (hydroxymethyl)phosphonate 91%
Dimethyl (hydroxymethyl)phosphonate

Precaución: for Research Use Only

Formula: C₃H₉O₄P
Boiling Pt: 209.4 °C (760 mm Hg)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD11053766
CAS Number: 24630-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR