Dimethyl (1,3-dihydro-3-oxo-1-isobenzofuranyl)phosphonate 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl (3-oxo-1,3-dihydroisobenzofuran-1-yl)phosphonate

A231847-100G A231847-10G A231847-250MG A231847-1G A231847-5G A231847-25G
103810-796EA 7 USD
103810-796 103850-550 76815-160 76815-162 76815-164 77089-448 77089-450 77303-924
Dimethyl (1,3-dihydro-3-oxo-1-isobenzofuranyl)phosphonate 98%
Dimethyl (1,3-dihydro-3-oxo-1-isobenzofuranyl)phosphonate

Precaución: for Research Use Only

Formula: C₁₀H₁₁O₅P
MW: 242.165 g/mol
Boiling Pt: 402.0±45.0 °C (760 mm Hg)
Melting Pt: 97…99 °C
Flash Pt: 210.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16883476
CAS Number: 61260-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR