Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate 97%

Supplier: Ambeed
Warning

Synonyms: Dimethyl (3,3-difluoro-2-oxoheptyl)phosphonate, (3,3-Difluoro-2-oxoheptyl)phosphonic acid Dimethyl ester

A110928-100MG A110928-1G A110928-5G
103797-756EA 56 USD
103797-756 103823-564 76821-818 77310-368 77310-370
Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate 97%
Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate

Precaución: for Research Use Only

Formula: C₉H₁₇F₂O₄P
MW: 258.20 g/mol
Boiling Pt: 301 °C (760 mm Hg)
Flash Pt: 135.8 °C
Storage Temperature: Ambient
MDL Number: MFCD11707108
CAS Number: 50889-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR