Dimethyl (2-oxopropyl)phosphonate 97%
Supplier: Ambeed
|
Warning
|
A181203-1G
A181203-25G
A181203-5G
A181203-100G
103807-586EA
5
USD
103807-586
103845-952
77083-470
77083-472
77083-474
77593-158
Dimethyl (2-oxopropyl)phosphonate 97%
Dimethyl (2-oxopropyl)phosphonate
Precaución: for Research Use Only
Formula:
C₅H₁₁O₄P MW: 166.12 g/mol Boiling Pt: 227 °C (760 mm Hg) Flash Pt: 93.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008769 CAS Number: 4202-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...