Diisopropyl succinate 98%

Supplier: Ambeed
Warning

Synonyms: Succinic Acid Diisopropyl Ester

A136412-1000G A136412-100G A136412-10G A136412-500G A136412-25G
76782-678EA 40 USD
76782-678 76782-680 76782-682 76782-684 77315-190
Diisopropyl succinate 98%
Diisopropyl succinate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 228 °C (760 mm Hg)
Melting Pt: 40 °C
Flash Pt: 96.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00059360
CAS Number: 924-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR