Diisopropyl phosphonate 97%
Supplier: Ambeed
|
Warning
|
A106505-100G
A106505-25G
103807-954EA
267
USD
103807-954
77107-676
77107-678
Diisopropyl phosphonate 97%
Diisopropyl phosphonate
Precaución: for Research Use Only
Formula:
C₆H₁₅O₃P Boiling Pt: 195.5 °C (760 mm Hg) Flash Pt: 69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00117905 CAS Number: 1809-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...