Diisopropyl Hydrazine-1,2-dicarboxylate 97%

Supplier: Ambeed
Danger

Synonyms: Diisopropyl hydrazine-1,2-dicarboxylate, Hydrazine-1,2-dicarboxylic acid Diisopropyl ester

A203934-1G A203934-10G A203934-5G A203934-250MG
103817-506EA 24 USD
103817-506 103848-166 77086-310 77164-684 77302-882 77683-572
Diisopropyl Hydrazine-1,2-dicarboxylate 97%
Diisopropyl Hydrazine-1,2-dicarboxylate

Precaución: For research use only.

Formula: C₈H₁₆N₂O₄
MW: 204.23 g/mol
Boiling Pt: 271.5 °C (760 mm Hg)
Flash Pt: 118 °C
Storage Temperature: Freezer
MDL Number: MFCD19443326
CAS Number: 19740-72-8
UN: 2930
ADR: 6.1,4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR