3-Oxo-cyclobutane-1,1-dicarboxylic acid diisopropyl ester 96%

Supplier: Ambeed
Warning

Synonyms: Diisopropyl 3-oxocyclobutane-1,1-dicarboxylate

A162069-100G A162069-10G A162069-500G A162069-1G A162069-25G
103805-564EA 5 USD
103805-564 103843-812 77150-926 77150-928 77150-930 77319-838 77319-840
3-Oxo-cyclobutane-1,1-dicarboxylic acid diisopropyl ester 96%
3-Oxo-cyclobutane-1,1-dicarboxylic acid diisopropyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₅
MW: 242.268 g/mol
Boiling Pt: 293.2±40.0 °C (760 mm Hg)
Flash Pt: 123.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06742778
CAS Number: 893724-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR