Tetrahydrothiophen-3-one 98%

Supplier: Ambeed
Warning

A219091-100G A219091-10G A219091-500G A219091-25G A219091-5G
103849-638EA 5 USD
103849-638 76785-910 76785-912 76785-914 77088-180 77303-522
Tetrahydrothiophen-3-one 98%
Tetrahydrothiophen-3-one

Precaución: for Research Use Only

Formula: C₄H₆OS
Boiling Pt: 175.2 °C (760 mm Hg)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00005412
CAS Number: 1003-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR