Dihydrofuran-3(2H)-one 97%

Supplier: Ambeed
Danger

A339141-25G A339141-5G A339141-10G A339141-1G A339141-250MG A339141-100G
103836-760EA 10 USD
103836-760 76839-780 76839-782 77167-502 77167-504 77167-506 77167-508 77612-942
Dihydrofuran-3(2H)-one 97%
Dihydrofuran-3(2H)-one

Precaución: for Research Use Only

Formula: C₄H₆O₂
Boiling Pt: 151.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07778393
CAS Number: 22929-52-8
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR