Glutaric anhydride 96%

Supplier: Ambeed
Warning

Synonyms: Dihydro-2H-pyran-2,6(3H)-dione, Dihydro-2H-pyran-2,6(3H)-dione

A677090-100G A677090-500G A677090-25G
103810-510EA 77 USD
103810-510 76806-574 76806-576 77308-112
Glutaric anhydride 96%
Glutaric anhydride

Precaución: for Research Use Only

Formula: C₅H₆O₃
MW: 114.1 g/mol
Boiling Pt: 287
Melting Pt: 50…55 °C
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00006679
CAS Number: 108-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR