Diflunisal 98%

Supplier: Ambeed
Warning

Synonyms: 2',4'-Difluoro-4-hydroxy[1,1'-biphenyl]-3-carboxylic acid, 2',4'-Difluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid

A193875-1G A193875-25G A193875-5G A193875-100G A193875-10G
103816-594EA 9 USD
103816-594 103847-202 77085-104 77085-106 77085-108 77097-174 77097-176
Diflunisal 98%
Diflunisal

Precaución: for Research Use Only

Formula: C₁₃H₈F₂O₃
MW: 250.1976 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057834
CAS Number: 22494-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR