Diethyl pyridine-3,5-dicarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Diethyl 3,5-Pyridinedicarboxylate, 3,5-Pyridinedicarboxylic acid diethyl ester

A168098-100G A168098-500G A168098-1G A168098-25G A168098-5G
103806-248EA 5 USD
103806-248 103844-506 76799-290 76799-292 77081-550 77081-552 77081-554
Diethyl pyridine-3,5-dicarboxylate 98%
Diethyl pyridine-3,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄
MW: 223.228 g/mol
Boiling Pt: 300.5 °C (760 mm Hg)
Flash Pt: 135.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00085755
CAS Number: 4591-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR