4-Oxoheptanedioc acid diethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Diethyl 4-oxoheptanedioate

A215314-100G A215314-10G A215314-500G A215314-25G A215314-5G A215314-1G
103818-476EA 5 USD
103818-476 103849-180 76800-742 76800-744 76800-746 77087-704 77087-706 77303-404
4-Oxoheptanedioc acid diethyl ester 98%
4-Oxoheptanedioc acid diethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₅
MW: 230.26 g/mol
Boiling Pt: 318.5 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009210
CAS Number: 6317-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR