Diethyl 3-hydroxycyclobutane-1,1-dicarboxylate 95%

Supplier: Ambeed
Warning

A329292-250MG A329292-5G
103825-524EA 168 USD
103825-524 103836-478 76817-138 77883-898
Diethyl 3-hydroxycyclobutane-1,1-dicarboxylate 95%
Diethyl 3-hydroxycyclobutane-1,1-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₅
Boiling Pt: 272.939 °C (760 mm Hg)
Flash Pt: 97.585 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11878435
CAS Number: 99974-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR