Diethyl-3,5-dimethyl-2,4-pyrroledicarboxylate 98%

Supplier: Ambeed
Warning

A179443-100G A179443-10G A179443-1G A179443-25G A179443-5G
103807-430EA 5 USD
103807-430 103845-776 76998-610 76998-612 77083-230 77083-232 77083-234
Diethyl-3,5-dimethyl-2,4-pyrroledicarboxylate 98%
Diethyl-3,5-dimethyl-2,4-pyrroledicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₄
Boiling Pt: 372.3 °C (760 mm Hg)
Melting Pt: 135…136 °C
Flash Pt: 179 °C
Storage Temperature: Ambient
MDL Number: MFCD00005218
CAS Number: 2436-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR