Diethyl propylmalonate 98%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2-propylmalonate

A178234-100G A178234-10G A178234-500G A178234-25G A178234-5G
103845-646EA 7 USD
103845-646 76799-602 76799-604 76799-606 77083-048 77083-050
Diethyl propylmalonate 98%
Diethyl propylmalonate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄
Boiling Pt: 221.5 °C (760 mm Hg)
Flash Pt: 91.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009168
CAS Number: 2163-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR