Diethyl methylmalonate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Diethyl 2-methylmalonate
A134379-100G
A134379-10G
A134379-500G
A134379-1000G
A134379-25G
103808-486EA
82
USD
103808-486
77190-138
77190-140
77190-142
77314-758
77314-760
Diethyl methylmalonate 97%
Diethyl methylmalonate
Precaución: for Research Use Only
Formula:
C₈H₁₄O₄ MW: 174.2 g/mol Boiling Pt: 204.4 °C (760 mm Hg) Flash Pt: 76.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009162 CAS Number: 609-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...