Diethyl methyloxalacetate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-Ethoxalylpropionate, 2-Ethoxalylpropionic Acid Ethyl Ester, Methyloxalacetic Acid Diethyl Ester, Diethyl 2-methyl-3-oxosuccinate
A124799-100G
A124799-10G
A124799-25G
A124799-500G
A124799-1G
A124799-5G
103800-576EA
6
USD
103800-576
77246-272
77246-274
77246-276
77246-278
77298-020
77312-720
Diethyl methyloxalacetate 95%
Diethyl methyloxalacetate
Precaución: for Research Use Only
Formula:
C₉H₁₄O₅ Boiling Pt: 249.9 °C (760 mm Hg) Flash Pt: 111.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009163 CAS Number: 759-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...