Diethyl 2-hydroxypentanedioate 97%

Supplier: Ambeed
Warning

A101939-100MG A101939-250MG A101939-1G
103796-210EA 485 USD
103796-210 77025-186 77025-188 77282-864
Diethyl 2-hydroxypentanedioate 97%
Diethyl 2-hydroxypentanedioate

Precaución: for Research Use Only

Formula: C₉H₁₆O₅
Boiling Pt: 291.8 °C (760 mm Hg)
Flash Pt: 107.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16620610
CAS Number: 69134-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR