(±)-Diethyl acetylsuccinate 97%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2-acetylsuccinate, Diethyl acetylsuccinate

A184287-10G A184287-1G A184287-25G A184287-5G
103815-738EA 5 USD
103815-738 103846-300 76799-828 77083-912 77083-914 77083-916
(±)-Diethyl acetylsuccinate 97%
(±)-Diethyl acetylsuccinate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₅
MW: 216.23 g/mol
Boiling Pt: 180…183 °C (50 mm Hg)
Melting Pt: 8 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009157
CAS Number: 1115-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR