Diethyl acetamidomalonate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Diethyl 2-acetamidomalonate
A262018-100G
A262018-500G
A262018-25G
103809-860EA
67
USD
103809-860
77250-164
77250-166
77304-844
Diethyl acetamidomalonate 95%
Diethyl acetamidomalonate
Precaución: for Research Use Only
Formula:
C₉H₁₅NO₅ MW: 217.22 g/mol Boiling Pt: 311.4 °C (760 mm Hg) Melting Pt: 93…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009146 CAS Number: 1068-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...