Diethyl 2,5-dihydroxyterephthalate 98%
Supplier: Ambeed
|
Warning
|
A355142-1G
A355142-25G
A355142-5G
A355142-100G
A355142-10G
103826-140EA
14
USD
103826-140
103837-144
76840-274
76840-276
76840-278
77015-754
77594-056
Diethyl 2,5-dihydroxyterephthalate 98%
Diethyl 2,5-dihydroxyterephthalate
Precaución: for Research Use Only
Formula:
C₁₂H₁₄O₆ MW: 254.24 g/mol Boiling Pt: 408.5 °C (760 mm Hg) Flash Pt: 155.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055430 CAS Number: 5870-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...