Diethyl-2,5-dibromoterephthalate 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dibromoterephthalic Acid Diethyl Ester, Diethyl 2,5-dibromoterephthalate

A302730-100G A302730-10G A302730-1G A302730-25G
103835-506EA 11 USD
103835-506 76827-896 76827-898 76827-900 76838-190
Diethyl-2,5-dibromoterephthalate 98%
Diethyl-2,5-dibromoterephthalate

Precaución: for Research Use Only

Formula: C₁₂H₁₂Br₂O₄
MW: 380.03 g/mol
Boiling Pt: 388.3 °C (760 mm Hg)
Flash Pt: 188.6 °C
Storage Temperature: Ambient
MDL Number: MFCD12407163
CAS Number: 18013-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR