Diethyl-2,5-dibromoadipate 98%

Supplier: Ambeed
Danger

Synonyms: Diethyl 2,5-dibromohexanedioate, 2,5-Dibromoadipic acid diethyl ester

A265909-5G A265909-10G A265909-1G A265909-500G
103852-660EA 6 USD
103852-660 77092-358 77113-804 77113-806 77113-808
Diethyl-2,5-dibromoadipate 98%
Diethyl-2,5-dibromoadipate

Precaución: for Research Use Only

Formula: C₁₀H₁₆Br₂O₄
MW: 360.04 g/mol
Melting Pt: 65…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075379
CAS Number: 869-10-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR