Diethyl iminodiacetate 97%

Supplier: Ambeed
Warning

Synonyms: Iminodiacetic Acid Diethyl Ester, Diethyl 2,2'-azanediyldiacetate

A237110-10G A237110-1G A237110-25G A237110-5G A237110-100G
103850-904EA 7 USD
103850-904 76771-306 76771-308 77089-966 77089-968 77568-292
Diethyl iminodiacetate 97%
Diethyl iminodiacetate

Precaución: For research use only.

Formula: C₈H₁₅NO₄
MW: 189.21 g/mol
Boiling Pt: 208 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00041925
CAS Number: 6290-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR