Diethyl 2-aminomalonate, N-BOC protected 97%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2-(tert-Butoxycarbonylamino)malonate, Diethyl 2-[(tert-butoxycarbonyl)amino]malonate, Diethyl 2-[(tert-butoxycarbonyl)amino]propane-1,3-dioate

A277724-250MG A277724-1G A277724-5G A277724-25G A277724-100G
103813-364EA 6 USD
103813-364 103853-300 76802-128 76802-130 77093-200 77093-202 77305-268 77879-517
Diethyl 2-aminomalonate, N-BOC protected 97%
Diethyl 2-aminomalonate, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₆
Boiling Pt: 218 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00239423
CAS Number: 102831-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR