Diethyl-2-(aminomethylene)malonate 97%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2-(aminomethylene)malonate

A171102-1G A171102-25G A171102-5G A171102-100G A171102-10G
103806-590EA 6 USD
103806-590 103844-864 77081-986 77081-988 77081-990 77191-410 77191-412
Diethyl-2-(aminomethylene)malonate 97%
Diethyl-2-(aminomethylene)malonate

Precaución: for Research Use Only

Formula: C₈H₁₃NO₄
MW: 187.20 g/mol
Boiling Pt: 263.4 °C (760 mm Hg)
Melting Pt: 56…59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075695
CAS Number: 6296-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR