Diethyl 1H-imidazole-4,5-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Bis(ethoxycarbonyl)-1H-imidazole

A147481-10G A147481-25G A147481-5G A147481-1G
103803-786EA 33 USD
103803-786 103841-890 77110-106 77299-586 77299-588 77317-262
Diethyl 1H-imidazole-4,5-dicarboxylate 97%
Diethyl 1H-imidazole-4,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₄
MW: 212.203 g/mol
Boiling Pt: 357.9 °C (760 mm Hg)
Melting Pt: 153…157 °C
Flash Pt: 170.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00723829
CAS Number: 1080-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR