Diethyl cyclopropane-1,1-dicarboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Diethyl 1,1-cyclopropanedicarboxylate, Diethyl 1,1-cyclopropane dicarboxyliate
A144505-100G
A144505-10G
A144505-500G
A144505-5G
A144505-25G
103841-488EA
5
USD
103841-488
76689-118
76689-120
76689-122
77299-390
77316-736
Diethyl cyclopropane-1,1-dicarboxylate 95%
Diethyl cyclopropane-1,1-dicarboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₄O₄ MW: 186.21 g/mol Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001283 CAS Number: 1559-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...