Diethyl-2-thienylmethylphosphonate 98%

Supplier: Ambeed
Warning

A109102-250MG A109102-5G A109102-1G
103797-446EA 29 USD
103797-446 76821-692 76821-694 77283-808
Diethyl-2-thienylmethylphosphonate 98%
Diethyl-2-thienylmethylphosphonate

Precaución: for Research Use Only

Formula: C₉H₁₅O₃PS
Boiling Pt: 325.7 °C (760 mm Hg)
Flash Pt: 150.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00128457
CAS Number: 2026-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR