Diethyl (hydroxymethyl)phosphonate 97% (by GC)

Supplier: Ambeed
Warning

A116997-100G A116997-10G A116997-25G A116997-500G A116997-1000G
103808-164EA 187 USD
103808-164 103824-690 76797-100 76797-102 76797-104 76797-106 77297-508
Diethyl (hydroxymethyl)phosphonate 97% (by GC)
Diethyl (hydroxymethyl)phosphonate

Precaución: for Research Use Only

Formula: C₅H₁₃O₄P
MW: 168.13 g/mol
Boiling Pt: 124…126 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00014418
CAS Number: 3084-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR